Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium acetate
Ammonium acetate 0.5% in aqueous solution
Ammonium acetate 0.5% in aqueous solution
Ammonium acetate 0.5% in aqueous solution
FFCHFF62585000
:
FF CHEMICALS
:
Ammonium acetate 0.5% in aqueous solution
FFCHFF62585000
:
FF CHEMICALS
:
Pk:
5 L
Seq:
5190
:
:
77.08 g/mol
:
Ambient
:
211-162-9
:
631-61-8
:
H₃CCOONH₄
:
MFCD00013066