Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium acetate
Ammonium acetate in aqueous solution
Ammonium acetate in aqueous solution
Ammonium acetate in aqueous solution
Supplier:
Ambion
Certificates
Expand image
Ammonium acetate in aqueous solution
Supplier:
Ambion
About this item
CAS Number:
631-61-8
:
Formula:
H₃CCOONH₄
MDL Number:
MFCD00013066
:
77.08 g/mol
Storage Temperature:
Ambient
EINECS:
211-162-9