Order Entry
Sweden
ContactUsLinkComponent
 
Acetic acid ≥99.8% (by GC) eluent additive, for LC-MS, Fluka™
Catalog # 49199-50ML
CAS Number:  
undefined
Acetic acid ≥99.8% (by GC) eluent additive, for LC-MS, Fluka™
Catalog # 49199-50ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Vapor Density:
    2.07 (vs air)
  • Assay:
    99.5 - 100.5 %
  • Purity (GC):
    Min. 99.8 %
  • Solidification Point:
    16.3 - 16.7 °C
  • Water (Karl Fischer):
    Max. 0.2 %
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.01 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.2 ppm
  • Cadmium (Cd):
    Max. 0.02 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.05 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Germanium (Ge):
    Max. 0.05 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.02 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.05 ppm
  • Lead (Pb):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.05 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.05 ppm
  • Zirconium (Zr):
    Max. 0.1 ppm
  • Chloride (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 0.5 ppm
  • Sulfate (SO₄):
    Max. 0.5 ppm
  • Suitability for the LC-MS:
    complying
  • Acetic aNHydride (GC):
    Max. 0.01 %
  • APHA:
    Max. 10
  • Explosion Limit:
    16 %, 92 °F
  • Refractive Index:
    n20/D 1.371 (lit.)

Specification Test Results

About this item

 :
 :60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
 :GHS02
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152