

Chloramine-T (sodium salt) 0.05 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF63769020
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
About this item
:
:227.65 g/mol
Storage Temperature:Ambient
EINECS:204-854-7
UN:3267
ADR:8,III
:GHS05
:GHS08
CAS Number:127-65-1
Formula:C₇H₇ClNNaO₂S
MDL Number:MFCD00000522