Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide 46% in aqueous solution
Potassium hydroxide 46% in aqueous solution
Potassium hydroxide 46% in aqueous solution
SWEDKA852602
:
SWED HANDLING
:
Potassium hydroxide 46% in aqueous solution
SWEDKA852602
:
SWED HANDLING
:
Pk:
1
Seq:
0001
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
1814
:
8,II
:
GHS05
:
1310-58-3
:
KOH
:
MFCD00003553