Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl orange
Methyl orange 0.05% in aqueous solution pH-indicator
Methyl orange 0.05% in aqueous solution pH-indicator
HACH14832
:
Hach
:
Methyl orange 0.05% in aqueous solution pH-indicator
HACH14832
:
Hach
:
Pack type:
Dropping bottle, marked
Pk:
1
Seq:
2440
Pk Info:
100 ml
:
:
327.34 g/mol
:
Ambient
:
208-925-3
:
547-58-0
:
C₁₄H₁₄N₃NaO₃S
:
MFCD00007502