Order Entry
Sweden
ContactUsLinkComponent
Barium chloride 0.025 mol/l (0.05 N) in aqueous solution
Barium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # FFCHFF64489010
Supplier:  FF CHEMICALS
CAS Number:  
Barium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # FFCHFF64489010
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
 :208.24 g/mol
Storage Temperature:Ambient
EINECS:233-788-1
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445