Order Entry
Sweden
ContactUsLinkComponent
Female pelvis model
Female pelvis model
Catalog # SOMMMS8/1
Supplier:  SOMMER MARCUS
undefined
Female pelvis model
Catalog # SOMMMS8/1
Supplier:  SOMMER MARCUS

Specifications

  • Weight:
    1,3 kg
  • Description:
    Female pelvis model
  • Material:
    SOMSO-Plast®
  • W×D×H:
    170×280×230 mm
  • Pk:
    1

Specifications

About this item

Female pelvis model separating in four parts:

  • Pelvic muscles and the muscles of pelvic floor
  • External and internal genital organs
  • Network of nerves and vessels

Median section and removable inner organs.

Weight: 1,3 kg