Warning signs, Sekuroka®
Made of PE film with colourfast print. Self- adhesive warning symbol labels.
- Resistant to water, oil and alkalis
- Temperature stable from −40 to +80 °C
- Suitable for indoor use
: According to DIN 4844, BGV/A8, ASR A1.3 and practice tested.