Industrial balances, Defender™ 3000 washdown
Leverantör: OHAUS
Certifikat
Om denna produkt
A range of modular scales for tough industrial environments. Easy to use scales that are ideal for general weighing and simple counting applications in production, packaging, and shipping and receiving areas.
- IP 66 stainless steel indicator
- Sturdy tubular platform design with high quality 304 stainless steel top plate
- Internal rechargeable battery