Screw caps with rubber liner, KIMBLE®
Leverantör: KIMBLEKONTES
Certifikat
Om denna produkt
Phenolic cap material and cemented-in rubber liner are resistant to the effects of repeated autoclaving. Designed from ASTM Specification E982.
Phenolic cap material and cemented-in rubber liner are resistant to the effects of repeated autoclaving. Designed from ASTM Specification E982.