Cut resistant gloves, Krytech 579
: MAPA
These seamless HDPE liners with PU coating provide standard protection for intricate handling in reasonably clean environments.
- Suitable for long-term wear: excellent abrasion resistance
- Excellent dexterity thanks to anatomical shape
- Ergonomic to reduce fatigue in the hand, with ventilated back
- Excellent tactile sensitivity due to the gloves reduced thickness
Ideal for precision and automotive assembly, the assembly of thin metal sheets, handling untrimmed parts or sheet metal, handling and sorting small, sharp parts, finishing work as well as for handling cardboard and paper.
: EN 388:2016 (4342B), ISO 13997: 5.3N (540 g); Cat. II
- Catalog No:
- MAPA34579089
- MAPA34579088
- MAPA34579086
- MAPA34579080
- MAPA34579081
- MAPA34579087
- Model:
- Krytech 579
- Krytech 579
- Krytech 579
- Krytech 579
- Krytech 579
- Krytech 579
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HDPE
- HDPE
- HDPE
- HDPE
- HDPE
- HDPE
- Finish:
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- 3/4 coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
Frequently Bought Together