Bottle Carrier
Catalog # DELT19700
Supplier: DELTALAB
Specifications
- For:6×1000 ml bottles
- Description:Bottle rack with folding handles, stainless steel
- W×D×H:380×240×130 mm
- Pk:1
Specifications
About this item
Bottle carrier made of stainless steel, with folding handles.
- Very good chemical resistance
- Very good temperature resistance
It is suitable for the transport of up to six (1 L) chemical reagent bottles.