0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Carboys
Carboys, narrow neck, LDPE, with tubulation
Carboys, narrow neck, LDPE, with tubulation
Carboys, narrow neck, LDPE, with tubulation
Supplier:
SEMADENI
Certificates
Expand image
Carboys, narrow neck, LDPE, with tubulation
Supplier:
SEMADENI
About this item
LDPE, translucent.
Good chemical resistance
Good temperature resistance
With silicone tubing and pinchcock