


Acetonitrile ≥99.9% (by GC), CHROMASOLV™ LC-MS Ultra tested for UHPLC-MS, tested for UHPLC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS08
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Explosion Limit:16 %
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 1 ppm
- Water (Karl Fischer):Max. 0.01 %
- Vapor Density:1.41 (vs air)
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0001 %
- Aluminium (Al):Max. 20 ppb
- Calcium (Ca):Max. 50 ppb
- Iron (Fe):Max. 20 ppb
- Potassium (K):Max. 50 ppb
- Magnesium (Mg):Max. 20 ppb
- Sodium (Na):Max. 50 ppb
- Lead (Pb):Max. 20 ppb
- Transmittance at 191 nm:Min. 30 %
- Transmittance at 195 nm:Min. 85 %
- Transmittance at 200 nm:Min. 97 %
- Transmittance at 215 nm:Min. 98 %
- Transmittance from 230 nm:Min. 99 %
- UHPLC-MS ESI(+) (Reserpine):Max. 5 ppb
- UHPLC-MS ESI(-) (Digoxin):Max. 20 ppb
- UHPLC Gradient drift 254 nm:Max. 3 mAU
- UHPLC Gradient peak 254 nm:Max. 0.6 mAU
- UHPLC Gradient drift 210 nm:Max. 8 mAU
- UHPLC Gradient peak 210 nm:Max. 2 mAU
- Suitability for the UHPLC-MS:complying
- Fluorescence (chinin) at 254 nm:Max. 0.3 ppb
- Fluorescence (chinin) at 365 nm:Max. 0.3 ppb
- APHA:Max. 10
- Refractive Index:n20/D 1.344 (lit.)