Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
Benzethonium chloride
 :  Merck
 
 
 
 
Benzethonium chloride
 :  Merck
 

 

  • Hill Formula:
    C₂₇H₄₂ClNO₂
  • Molar Mass:
    448.08 g/mol
  • Ignition temperature:
    380 °C
  • Melting Point:
    160 - 166 °C
  • Vapor pressure:
     - 0.001 hPa (25 °C)
  • Bulk density:
    440 kg/m³
  • Solubility:
    (20 °C) soluble
  • LD 50 oral:
    LD50 Rat 295 mg/kg
  • Signal Word:
    Danger
  • Storage class:
    6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
  • WGK:
    WGK 2 water endangering
  • R Phrase:
    R 22-34-50/53
  • S Phrase:
    S 26-36/37/39-45-61