Linalyl acetate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:115-95-7
:
Formula:C₁₂H₂₀O₂
MDL Number:MFCD00008907
:196.2860 g/mol
Merck Index:14,05496
Boiling Pt:220°C
Flash Pt:85°C
EINECS:204-116-4
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %