Order Entry
Sweden
ContactUsLinkComponent
Linalyl acetate
Linalyl acetate
Catalog # EHERC10016230
Supplier:  EHRENSTORFER
CAS Number:  
Linalyl acetate
Catalog # EHERC10016230
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :196.2860 g/mol
EINECS:204-116-4
CAS Number:115-95-7
Formula:C₁₂H₂₀O₂
MDL Number:MFCD00008907