(R)-(+)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:147081-49-0
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD03427030
:186.25 g/mol
Flash Pt:91°C
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 97.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %