Quartz cuvettes for spectrophotometers
Catalog # WAPR80-2103-68
Supplier: Biochrom
Specifications
- Path length:5 mm
- Type:Ultra micro
- Volume:5 – 7 µl
- Description:Quartz cuvettes
- Seq:010
- Pk:1
Specifications
About this item
Life science cuvettes for methods with rapid scanning, such as nucleic acid quantification, proteins and cell density.