Magnetic stirring bar set
Magnetic stirring bar set
Catalog # AZLOSWN515
Supplier: SciLabware
Jump to:
Product Details & Documents
PTFE covered stirring bars, useful assortment of the most popular types and sizes.
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Ordering information: Supplied in a red polypropylene compartmented box with opaque natural lid. Set contains: Two each of 12×4,5; 25×6; 45×8 and 60×9,5 mm pivot ring type, two each of 20×10; 25×12; 35×16 and 50×20 mm oval type and three each of 7×2 and 10×3 mm micro stirrers.
Specifications
- Description:Magnetic stirring bar set