Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Desoxyribonuclease I
Desoxyribonuclease I
Desoxyribonuclease I
MSPPLK003172
:
MARKET SOURCE PART PROCESS
:
Desoxyribonuclease I
MSPPLK003172
:
MARKET SOURCE PART PROCESS
:
Pk:
5 Vial
CAS No.:
9003-98-9
Storage conditions:
2...8 °C
Form:
Lyophilised
Enzyme name:
Desoxyribonuclease I
Enzyme activity:
≥1000 units/vial
Enzyme synonyms:
DNase I