Order Entry
Sweden
ContactUsLinkComponent
2-Ethylhexyl salicylate
2-Ethylhexyl salicylate
Catalog # EHERC16903520
Supplier:  EHRENSTORFER
CAS Number:  
2-Ethylhexyl salicylate
Catalog # EHERC16903520
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :250.34 g/mol
EINECS:204-263-4
CAS Number:118-60-5
Formula:C₁₅H₂₂O₃
MDL Number:MFCD00053300