Cholecalciferol (Vitamin D3) ≥98.0% (by HPLC)
Catalog # TCIAC0314-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+103 to +112 deg (C=0.5, EtOH)
- Absorbance:0.450 to 0.510 (10mg/L, EtOH, 263.0 to 267.0 nm)
Specification Test Results
About this item
:
:384.65 g/mol
Merck Index:14,10019
Storage Temperature:Freezer
EINECS:200-673-2
UN:2811
ADR:6.1,II
:GHS06
:GHS08
CAS Number:67-97-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131