Order Entry
Sweden
ContactUsLinkComponent
(4-Nitrophenyl)-β-D-glucopyranoside ≥98.0% (by HPLC) substrate for beta-D-Glucosidase
(4-Nitrophenyl)-β-D-glucopyranoside ≥98.0% (by HPLC) substrate for beta-D-Glucosidase
Supplier:  TCI
Certificates
undefined
(4-Nitrophenyl)-β-D-glucopyranoside ≥98.0% (by HPLC) substrate for beta-D-Glucosidase
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    164.0 to 168.0 deg-C
  • Specific rotation [a]20/D:
    -98.0 to -105.0 deg (C=1, H2O)
  • Water:
    3.8 to 7.0 %

Specification Test Results

Related Information

Frequently Bought Together