0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
2,4-Difluoropyridine
2,4-Difluoropyridine ≥98.0% (by GC)
2,4-Difluoropyridine ≥98.0% (by GC)
:
TCI
2,4-Difluoropyridine ≥98.0% (by GC)
:
TCI
:
34941-90-7
:
:
C₅H₃F₂N
:
MFCD04114162
:
106°C
:
1993
:
3,III
:
115.08 g/mol
:
GHS02
:
GHS07
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 98.0 %
Manual Fluorination Reagents, Fluorinated Building Blocks