Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Lactose

Glass bottle for solids, 500 g
Catalog # COND1902
Supplier:   LABORATORIOS CONDA
Lactose
Catalog # COND1902
Supplier:  LABORATORIOS CONDA
CAS Number:  63-42-3
Lactose

Glass bottle for solids, 500 g

CAS Number:  63-42-3
Price
680,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Milk sugar, D-Lactose, Anhydrous Lactose, (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal
  • CAS Number:63-42-3
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00151251
  • Molecular Weight:342.3 g/mol
  • Merck Index:12,05356
  • Boiling Pt:592 °C (1013 hPa)
  • Melting Pt:222.8 °C
  • Storage Temperature:Ambient
  • EINECS:200-559-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications