
2,4-Dimethoxytoluene, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:38064-90-3
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00010749
:152.1904 g/mol
Boiling Pt:110...120 °C/30 mmHg (lit.)
Density:1,036 g/ml (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Infrared spectrum:Conforms to Structure
- Purity (GC):≥98.5 %