
Ionomycin calcium salt ≥99%
Catalog # J60628.X0
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:5 mg
- Seq:2210
Specifications
About this item
:
:747.1 g/mol
:GHS07
Storage Temperature:Frys
CAS Number:56092-82-1
Formula:C₄₁H₇₀CaO₉
MDL Number:MFCD00151183