
Ionomycin calcium salt ionophore
Catalog # 59007.
Supplier: Biotium
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:1 mg
- Seq:2070
Specifications
About this item
:
:747.1 g/mol
:GHS07
Storage Temperature:Kylskåp
CAS Number:56092-82-1
Formula:C₄₁H₇₀CaO₉
MDL Number:MFCD00151183