D-(-)-Luciferin ≥98.0% (by HPLC) chemiluminescence reagent
: TCI
:2591-17-5
:
:C₁₁H₈N₂O₃S₂
:MFCD00042929
:280.3228 g/mol
:14,04086
:186°C
:Freezer
:219-981-3
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 90.0 %
- Specific rotation [a]20/D:-30.0 to -34.0 deg (C=1, DMF)
- Luminescent test:to pass test
Frequently Bought Together