

Methylmercury hydroxide 1 M in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1184-57-2
:
Formula:CH₄HgO
MDL Number:MFCD00013924
:232.632 g/mol
Storage Temperature:Ambient
EINECS:214-667-2
UN:2024
ADR:6.1,II
:GHS06
:GHS08
Related Information
Frequently Bought Together