Order Entry
Sweden
ContactUsLinkComponent
Pesticide reference standard, cis-Chlordane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, cis-Chlordane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11201000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, cis-Chlordane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11201000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    5103-71-9
  • Description:
    cis-Chlordane 100 µg/ml in cyclohexane
  • Formula:
    C₁₀H₆Cl₈
  • Description:
    cis-Chlordane

Specifications

About this item