Filter holders, Swinnex®
Supplier: Merck
Certificates
About this item
These filter holders are used to ultraclean or sterilise small volumes of liquids dispensed by syringes.
47 mm filter holder connected with tubing to pressurised reservoir or automatic filling machine for filtering larger volumes.
Specifications
- Catalog No:
- 511-0852
- 511-0853
- 511-0851
- Filtration area:
- 3,4 cm²
- 13,8 cm²
- 0,7 cm²
- Inlet/outlet connections:
- Female Luer lock®/male luer slip
- 6,35 mm NPTM /6,35 mm hose connector, with interior female luer slip
- Female Luer lock®/male luer slip