Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nystatin
Nystatin
Nystatin
Nystatin
Leverantör:
Thermo Fisher Scientific
Certifikat
Expand image
Nystatin
Leverantör:
Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:
1400-61-9
:
Formel:
C₄₇H₇₅NO₁₇
MDL-nummer:
MFCD25541651
:
926.09 g/mol
Merck Index:
14,6736
Smält Pt:
160 °C
EINECS:
215-749-0
:
GHS07
Förvaringstemperatur:
Freezer
Relaterad information
Documentation
Certifikat
Sök efter LOT-nummer
SDS
msds_ALFAJ62486
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation