Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate
Leverantör:
Thermo Fisher Scientific
Certifikat
Expand image
2'-Deoxyguanosine monohydrate
Leverantör:
Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:
312693-72-4
:
Formel:
C₁₀H₁₃N₅O₄·H₂O
MDL-nummer:
MFCD00150760
:
285.26 g/mol
Förvaringstemperatur:
Ambient
EINECS:
213-505-8
Relaterad information
Documentation
Certifikat
Sök efter LOT-nummer
SDS
msds_ALFAJ60741
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation