Order Entry
Sweden
ContactUsLinkComponent
DEAE-Dextran, powder
DEAE-Dextran, powder
Catalog # J63781.14
CAS Number:  
DEAE-Dextran, powder
Catalog # J63781.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Specification Test Results

  • Form:
    Powder
  • Free DEAE-Cl:
    <20 ppm
  • Loss on Drying:
    <10%
  • Molecular Weight (viscosity calc.):
    500,000 ± 20%
  • Total Nitrogen:
    3.0 - 3.8%
  • Optical purity:
    NMT 0.2 absorption at 375 nm
  • pH (aqueous solution):
    5.0 to 7.0
  • Sulfated Ash:
    <2%
  • Viscosity:
    0.4 to 0.6

Specification Test Results

About this item

CAS Number:9015-73-0
 :
MDL Number:MFCD00081561
Merck Index:14,2936
Storage Temperature:Ambient