Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-2-methylphenylboronic acid
4-Fluoro-2-methylphenylboronic acid ≥98%
4-Fluoro-2-methylphenylboronic acid ≥98%
:
Thermo Fisher Scientific
4-Fluoro-2-methylphenylboronic acid ≥98%
:
Thermo Fisher Scientific
:
139911-29-8
:
MFCD17214439
:
C₇H₈BFO₂
:
MFCD02093072
:
153.95 g/mol
:
199…201 °C
:
GHS07
:
Ambient
msds_ALFAB24117
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years