
2-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:352535-96-7
:
Formula:C₇H₅BF₄O₂
MDL Number:MFCD03701538
:207.92 g/mol
Melting Pt:109°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):98.0 to 110.0 %