3,4,5-Tribromo-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17635-44-8
:
Formula:C₃HBr₃N₂
MDL Number:MFCD00040248
:304.78 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:186.0 to 190.0 deg-C