PCR Plates for thermal cyclers
Supplier: Thermo Fisher Scientific
Certificates
About this item
Piko® PCR plates utilise ultra-thin wall (UTW) technology to dramatically reduce the thickness of the well walls, and are specifically designed for use in the Piko® thermal cycler and the PikoReal® Real Time PCR system. Piko® PCR plates have the footprint of a microscope slide. Four Piko® 24-well (96-well) plates can be snapped into the plate frame to create a standard 96-well (384-well) plate.
- Ultra-thin wall (UTW) for fast PCR/qPCR applications
- Low profile
- Compatible with standard multi-channel pipettes and liquid handling platforms
- Well spacing and footprint conform to industry dimensions
These plates are made of PP.