Order Entry
Sweden
ContactUsLinkComponent
Haematocrit sealing material
Haematocrit sealing material
Katalog # HECH40566010
undefined
Haematocrit sealing material
Katalog # HECH40566010
Specifications
  • Description:
    Hematocrit sealing wax, non-drying, in plates of polystyrene with numbered positions, for sealing hematocrit tubes
  • Pk:
    1
Specifications
About this item
This sealing material prevents the draining of blood samples. For sealing several hundred micro hematocrit capillaries.