Ponceau S ≥85.0% (by total nitrogen basis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:6226-79-5
:
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892
:760.56 g/mol
EINECS:228-319-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (with Total Nitrogen):min. 85.0 %
- Protein staining test:to pass test