Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pig Pancreatin (from Pancreas)
Pig Pancreatin (from Pancreas)
:
TCI
Pig Pancreatin (from Pancreas)
:
TCI
CAS No.:
8049-47-6
Source:
Pancreas
Species:
Pig
Storage conditions:
Refrigerator
Enzyme name:
Pancreatin
Enzyme activity:
4xJP
Manual Bioscience and Analyticalscience