1-Chloro-2-butene (mixture of cis and trans isomers) cis- and trans-mixture ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:591-97-9
:
Formula:C₄H₇Cl
MDL Number:MFCD00000987
:90.55 g/mol
Merck Index:14,02130
Boiling Pt:83°C
Density:0,9491 g/cm³ (0 °C)
Flash Pt:–19°C
EINECS:209-739-5
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 % (total of isomer)