2-Phenylethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:60-12-8
:
Formula:C₆H₅CH₂CH₂OH
MDL Number:MFCD00002886
:122.17 g/mol
Merck Index:14,07224
Boiling Pt:219°C
Melting Pt:103…105 °C
Density:1,02 g/cm³ (20 °C)
Flash Pt:108°C
Storage Temperature:Ambient
EINECS:200-456-2
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %