1-Bromo-9,10-anthraquinone ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:632-83-7
:
Formula:C₁₄H₇BrO₂
MDL Number:MFCD00451689
:287.108 g/mol
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:188.0 to 192.0 deg-C