Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiophene-3-methylamine
Thiophene-3-methylamine
Thiophene-3-methylamine
Thiophene-3-methylamine
Supplier:
Thermo Scientific
Certificates
Expand image
Thiophene-3-methylamine
Supplier:
Thermo Scientific
About this item
CAS Number:
27757-86-4
:
Formula:
C₅H₇NS
MDL Number:
MFCD01529872
:
113.18 g/mol