2-Acetamido-5-nitrothiazole
Catalog # EHERC10012500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:187.18 g/mol
EINECS:205-414-7
CAS Number:140-40-9
Formula:C₅H₅N₃O₃S
MDL Number:MFCD00022438