Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tricaine methanesulphonate
Tricaine methanesulphonate
Tricaine methanesulphonate
Tricaine methanesulphonate
Supplier:
RESEARCH ORGANICS
Certificates
Expand image
Tricaine methanesulphonate
Supplier:
RESEARCH ORGANICS
About this item
CAS Number:
886-86-2
:
Formula:
C₁₀H₁₅NO₅S
MDL Number:
MFCD00013176
:
261.29 g/mol
EINECS:
212-956-8